Assay Details
| Gene Symbol: sc5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004630.1 | NP_001004630.1 | 4 - 5 | 497 | 77 | |
| GenBank mRNA | BC081395.1 | - | 4 - 5 | 497 | 77 | |
More Information
Set Membership: |
|

sc5d
Genomic Map