Assay Details
Gene Symbol: cnot3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005158349.4 | XP_005158406.1 | 11 - 12 | 1979 | 64 | |
GenBank mRNA | BC081678.1 | - | 8 - 9 | 1025 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
