Assay Details
| Gene Symbol: plp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005586.2 | NP_001005586.1 | 6 - 7 | 885 | 76 | |
| XM_005165122.4 | XP_005165179.1 | 7 - 8 | 905 | 76 | ||
| XM_005165123.4 | XP_005165180.1 | 7 - 8 | 902 | 76 | ||
| XM_005165124.4 | XP_005165181.1 | 6 - 7 | 794 | 76 | ||
| GenBank mRNA | AY070260.1 | - | 6 - 7 | 726 | 76 | |
| AY070262.1 | - | 7 - 8 | 828 | 76 | ||
| AY070263.1 | - | 6 - 7 | 885 | 76 | ||
| BC081573.1 | - | 6 - 7 | 730 | 76 | ||
More Information
Set Membership: |
|

plp1b
Genomic Map