Assay Details
| Gene Symbol: cox6a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005592.1 | NP_001005592.1 | 2 - 3 | 253 | 98 | |
| GenBank mRNA | BC081530.1 | - | 2 - 3 | 253 | 98 | |
| BC164950.1 | - | 2 - 3 | 274 | 98 | ||
More Information
Set Membership: |
|

cox6a1
Genomic Map