Assay Details
| Gene Symbol: myl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005955.1 | NP_001005955.1 | 3 - 4 | 234 | 96 | |
| XM_005166077.4 | XP_005166134.1 | 3 - 4 | 313 | 96 | ||
| XM_005166078.4 | XP_005166135.1 | 3 - 4 | 312 | 96 | ||
| GenBank mRNA | BC083444.1 | - | 3 - 4 | 234 | 96 | |
| BC152157.1 | - | 3 - 4 | 234 | 96 | ||
More Information
Set Membership: |
|

myl6
Genomic Map