Assay Details
Gene Symbol: gemin8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007317.1 | NP_001007318.1 | 2 - 3 | 645 | 66 | |
XM_005168381.4 | XP_005168438.1 | 3 - 4 | 548 | 66 | ||
GenBank mRNA | BC085647.1 | - | 2 - 3 | 645 | 66 | |
BC129489.1 | - | 3 - 4 | 609 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
