Assay Details
| Gene Symbol: slc25a44a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007350.1 | NP_001007351.1 | 3 - 4 | 986 | 98 | |
| GenBank mRNA | BC085527.1 | - | 3 - 4 | 986 | 98 | |
More Information
Set Membership: |
|

slc25a44a
Genomic Map