Assay Details
| Gene Symbol: grb2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007769.1 | NP_001007770.1 | 2 - 3 | 572 | 100 | |
| GenBank mRNA | BC085549.1 | - | 2 - 3 | 572 | 100 | |
| BC165040.1 | - | 1 - 2 | 106 | 100 | ||
More Information
Set Membership: |
|

grb2a
Genomic Map