Assay Details
| Gene Symbol: dhodh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008608.1 | NP_001008608.1 | 5 - 6 | 780 | 89 | |
| XM_005166562.3 | XP_005166619.1 | 5 - 6 | 758 | 89 | ||
| GenBank mRNA | BC086819.1 | - | 5 - 6 | 780 | 89 | |
| BC164266.1 | - | 5 - 6 | 731 | 89 | ||
More Information
Set Membership: |
|

dhodh
Genomic Map