Assay Details
| Gene Symbol: mcamb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012367.2 | NP_001012367.2 | 13 - 14 | 1707 | 116 | |
| GenBank mRNA | AY497061.1 | - | 13 - 14 | 1686 | 116 | |
| AY746594.1 | - | 13 - 14 | 1707 | 116 | ||
| BC162589.1 | - | 13 - 14 | 1699 | 116 | ||
More Information
Set Membership: |
|

mcamb
Genomic Map