Assay Details
| Gene Symbol: mcamb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012367.2 | NP_001012367.2 | 2 - 3 | 261 | 127 | |
| XM_005157570.2 | XP_005157627.1 | 2 - 3 | 127 | |||
| XM_021481353.1 | XP_021337028.1 | 2 - 3 | 127 | |||
| GenBank mRNA | AY746594.1 | - | 2 - 3 | 261 | 127 | |
| BC162589.1 | - | 2 - 3 | 127 | |||
More Information
Set Membership: |
|

mcamb
Genomic Map