Assay Details
| Gene Symbol: ccdc115 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013295.2 | NP_001013313.1 | 3 - 4 | 442 | 121 | |
| XM_009296733.3 | XP_009295008.1 | 4 - 5 | 499 | 121 | ||
| GenBank mRNA | BC090281.1 | - | 3 - 4 | 446 | 121 | |
More Information
Set Membership: |
|

ccdc115
Genomic Map