Assay Details
Gene Symbol: erc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015065.1 | NP_001015065.1 | 3 - 4 | 1147 | 68 | |
XM_009298003.3 | XP_009296278.1 | 4 - 5 | 1492 | 68 | ||
XM_009298005.3 | XP_009296280.1 | 4 - 5 | 1493 | 68 | ||
XM_009298006.3 | XP_009296281.1 | 4 - 5 | 1491 | 68 | ||
XM_021470486.1 | XP_021326161.1 | 3 - 4 | 1222 | 68 | ||
XM_021470487.1 | XP_021326162.1 | 3 - 4 | 1141 | 68 | ||
GenBank mRNA | AY916778.1 | - | 3 - 4 | 1147 | 68 | |
BC171689.1 | - | 3 - 4 | 1222 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
