Assay Details
Gene Symbol: erc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015065.1 | NP_001015065.1 | 9 - 10 | 2061 | 60 | |
XM_009298003.3 | XP_009296278.1 | 10 - 11 | 2406 | 60 | ||
XM_009298005.3 | XP_009296280.1 | 10 - 11 | 2407 | 60 | ||
XM_009298006.3 | XP_009296281.1 | 10 - 11 | 2405 | 60 | ||
XM_021470486.1 | XP_021326161.1 | 9 - 10 | 2136 | 60 | ||
XM_021470487.1 | XP_021326162.1 | 9 - 10 | 2055 | 60 | ||
GenBank mRNA | AY916778.1 | - | 9 - 10 | 2061 | 60 | |
BC171689.1 | - | 9 - 10 | 2136 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
