Assay Details
Gene Symbol: cacng8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001326483.1 | NP_001313412.1 | 3 - 4 | 604 | 90 | |
XM_005159434.4 | XP_005159491.1 | 3 - 4 | 628 | 90 | ||
XM_009294042.3 | XP_009292317.1 | 3 - 4 | 707 | 90 | ||
GenBank mRNA | BC093221.1 | - | 3 - 4 | 604 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
