Assay Details
| Gene Symbol: ndufb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017880.1 | NP_001017880.1 | 2 - 3 | 288 | 99 | |
| GenBank mRNA | BC092968.1 | - | 2 - 3 | 288 | 99 | |
| BC154160.1 | - | 2 - 3 | 290 | 99 | ||
More Information
Set Membership: |
|

ndufb9
Genomic Map