Assay Details
Gene Symbol: tbc1d20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025172.1 | NP_001020343.1 | 1 - 2 | 65 | 73 | |
GenBank mRNA | BC096811.1 | - | 1 - 2 | 65 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
