Assay Details
Gene Symbol: dusp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025177.1 | NP_001020348.1 | 2 - 3 | 303 | 66 | |
XM_005163623.4 | XP_005163680.1 | 2 - 3 | 344 | 66 | ||
GenBank mRNA | BC097131.1 | - | 2 - 3 | 303 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
