Assay Details
| Gene Symbol: ilf3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030065.1 | NP_001025236.1 | 18 - 19 | 2460 | 107 | |
| XM_005165723.4 | XP_005165780.1 | 18 - 19 | 2501 | 107 | ||
| XM_009302153.3 | XP_009300428.1 | 18 - 19 | 2504 | 107 | ||
| GenBank mRNA | BC096956.1 | - | 18 - 19 | 2460 | 107 | |
More Information
Set Membership: |
|

ilf3a
Genomic Map