Assay Details
| Gene Symbol: ilf3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030065.1 | NP_001025236.1 | 12 - 13 | 1496 | 109 | |
| XM_009302153.3 | XP_009300428.1 | 12 - 13 | 1540 | 109 | ||
| GenBank mRNA | BC096956.1 | - | 12 - 13 | 1496 | 109 | |
More Information
Set Membership: |
|

ilf3a
Genomic Map