Assay Details
Gene Symbol: psph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033099.1 | NP_001028271.1 | 5 - 6 | 618 | 88 | |
GenBank mRNA | BC100056.1 | - | 5 - 6 | 618 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
