Assay Details
Gene Symbol: nr6a1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033720.1 | NP_001028892.1 | 9 - 10 | 1338 | 65 | |
GenBank mRNA | BC097237.1 | - | 9 - 10 | 1338 | 65 | |
DQ017610.1 | - | 7 - 8 | 1151 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
