Assay Details
| Gene Symbol: pi4kaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034972.1 | NP_001030144.1 | 12 - 13 | 1421 | 104 | |
| XM_005172596.4 | XP_005172653.1 | 13 - 14 | 1649 | 104 | ||
| XM_009305081.3 | XP_009303356.1 | 13 - 14 | 1650 | 104 | ||
| GenBank mRNA | AY929292.1 | - | 12 - 13 | 1421 | 104 | |
More Information
Set Membership: |
|

pi4kaa
Genomic Map