Assay Details
| Gene Symbol: antxr2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044709.2 | NP_001038174.2 | 14 - 15 | 1502 | 65 | |
| GenBank mRNA | BC139636.1 | - | 14 - 15 | 1502 | 65 | |
| DQ415957.1 | - | 14 - 15 | 1157 | 65 | ||
More Information
Set Membership: |
|

antxr2a
Genomic Map