Assay Details
| Gene Symbol: antxr2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044709.2 | NP_001038174.2 | 3 - 4 | 618 | 116 | |
| XM_005165319.4 | XP_005165376.1 | 3 - 4 | 641 | 116 | ||
| GenBank mRNA | BC139636.1 | - | 3 - 4 | 618 | 116 | |
| DQ415957.1 | - | 3 - 4 | 273 | 116 | ||
More Information
Set Membership: |
|

antxr2a
Genomic Map