Assay Details
Gene Symbol: kcnh5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276280.1 | NP_001263209.1 | 11 - 12 | 2192 | 81 | |
GenBank mRNA | BC163622.1 | - | 11 - 12 | 2192 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
