Assay Details
| Gene Symbol: npr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044937.2 | NP_001038402.1 | 14 - 15 | 2269 | 75 | |
| XM_005170050.4 | XP_005170107.1 | 15 - 16 | 2949 | 75 | ||
More Information
Set Membership: |
|

npr1a
Genomic Map