Assay Details
| Gene Symbol: gna11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045036.2 | NP_001038501.1 | 3 - 4 | 460 | 83 | |
| XM_005170475.4 | XP_005170532.1 | 3 - 4 | 466 | 83 | ||
More Information
Set Membership: |
|

gna11a
Genomic Map