Assay Details
| Gene Symbol: acsl3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045128.1 | NP_001038593.1 | 10 - 11 | 1514 | 66 | |
| XM_005159236.4 | XP_005159293.1 | 12 - 13 | 2022 | 66 | ||
More Information
Set Membership: |
|

acsl3a
Genomic Map