Assay Details
| Gene Symbol: acsl3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045128.1 | NP_001038593.1 | 13 - 14 | 1978 | 89 | |
| XM_005159236.4 | XP_005159293.1 | 15 - 16 | 2486 | 89 | ||
More Information
Set Membership: |
|

acsl3a
Genomic Map