Assay Details
| Gene Symbol: acsl3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045128.1 | NP_001038593.1 | 1 - 2 | 361 | 72 | |
| XM_005159236.4 | XP_005159293.1 | 3 - 4 | 869 | 72 | ||
More Information
Set Membership: |
|

acsl3a
Genomic Map