Assay Details
Gene Symbol: scn8ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045183.1 | NP_001038648.1 | 11 - 12 | 1914 | 83 | |
XM_005166072.3 | XP_005166129.1 | 12 - 13 | 2188 | 83 | ||
XM_009302566.2 | XP_009300841.1 | 12 - 13 | 2187 | 83 | ||
GenBank mRNA | DQ149509.1 | - | 11 - 12 | 1914 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
