Assay Details
Gene Symbol: scn8ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045183.1 | NP_001038648.1 | 1 - 2 | 273 | 93 | |
XM_005166071.3 | XP_005166128.1 | 2 - 3 | 547 | 93 | ||
XM_005166072.3 | XP_005166129.1 | 2 - 3 | 547 | 93 | ||
XM_009302566.2 | XP_009300841.1 | 2 - 3 | 546 | 93 | ||
XM_021476941.1 | XP_021332616.1 | 2 - 3 | 548 | 93 | ||
XM_021476942.1 | XP_021332617.1 | 2 - 3 | 1934 | 93 | ||
GenBank mRNA | DQ149509.1 | - | 1 - 2 | 273 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
