Assay Details
Gene Symbol: scn8ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045183.1 | NP_001038648.1 | 2 - 3 | 406 | 129 | |
GenBank mRNA | DQ149509.1 | - | 2 - 3 | 406 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
