Assay Details
| Gene Symbol: ppp1r15b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045219.2 | NP_001038684.3 | 2 - 3 | 1474 | 76 | |
| GenBank mRNA | BC134978.1 | - | 2 - 3 | 1474 | 76 | |
More Information
Set Membership: |
|

ppp1r15b
Genomic Map