Assay Details
Gene Symbol: acsf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045419.2 | NP_001038884.2 | 8 - 9 | 1062 | 58 | |
NM_001139438.1 | NP_001132910.1 | 7 - 8 | 1222 | 58 | ||
GenBank mRNA | BC122098.1 | - | 8 - 9 | 1057 | 58 | |
CF348078.1 | - | 4 - 5 | 444 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
