Assay Details
Gene Symbol: sgk2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076564.1 | NP_001070032.1 | 4 - 5 | 265 | 91 | |
XM_009292340.3 | XP_009290615.1 | 4 - 5 | 334 | 91 | ||
GenBank mRNA | BC124529.1 | - | 4 - 5 | 265 | 91 | |
BC164500.1 | - | 4 - 5 | 249 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
