Assay Details
Gene Symbol: smyd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076594.1 | NP_001070062.1 | 2 - 3 | 358 | 70 | |
XM_021479202.1 | XP_021334877.1 | 2 - 3 | 479 | 70 | ||
GenBank mRNA | BC124341.1 | - | 2 - 3 | 358 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
