Assay Details
| Gene Symbol: smap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077778.1 | NP_001071246.1 | 7 - 8 | 860 | 53 | |
| XM_021480668.1 | XP_021336343.1 | 8 - 9 | 971 | 53 | ||
| XM_021480669.1 | XP_021336344.1 | 8 - 9 | 968 | 53 | ||
| XM_021480670.1 | XP_021336345.1 | 7 - 8 | 895 | 53 | ||
| GenBank mRNA | BC124755.1 | - | 7 - 8 | 860 | 53 | |
| BC165724.1 | - | 7 - 8 | 736 | 53 | ||
More Information
Set Membership: |
|

smap1
Genomic Map