Assay Details
Gene Symbol: kaznb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080082.2 | NP_001073551.2 | 2 - 3 | 652 | 76 | |
XM_009306043.3 | XP_009304318.2 | 5 - 6 | 810 | 76 | ||
XM_017358249.2 | XP_017213738.1 | 5 - 6 | 821 | 76 | ||
XM_017358250.2 | XP_017213739.1 | 5 - 6 | 810 | 76 | ||
XM_021479663.1 | XP_021335338.1 | 2 - 3 | 322 | 76 | ||
GenBank mRNA | BC128836.1 | - | 2 - 3 | 487 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
