Assay Details
| Gene Symbol: nfkbie | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080089.1 | NP_001073558.1 | 4 - 5 | 968 | 62 | |
| GenBank mRNA | BC128831.1 | - | 4 - 5 | 968 | 62 | |
More Information
Set Membership: |
|

nfkbie
Genomic Map