Assay Details
| Gene Symbol: isy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083563.2 | NP_001077032.1 | 3 - 4 | 324 | 73 | |
| GenBank mRNA | BC115265.1 | - | 3 - 4 | 98 | 73 | |
More Information
Set Membership: |
|

isy1
Genomic Map