Assay Details
| Gene Symbol: acbd5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100088.1 | NP_001093558.1 | 11 - 12 | 1189 | 90 | |
| XM_005171138.3 | XP_005171195.1 | 11 - 12 | 1240 | 90 | ||
| XM_005171140.2 | XP_005171197.1 | 11 - 12 | 1219 | 90 | ||
| XM_021478275.1 | XP_021333950.1 | 11 - 12 | 1219 | 90 | ||
| XM_021478280.1 | XP_021333955.1 | 10 - 11 | 1023 | 90 | ||
More Information
Set Membership: |
|

acbd5b
Genomic Map