Assay Details
Gene Symbol: hoxa10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131155.2 | NP_571230.2 | 1 - 2 | 865 | 62 | |
GenBank mRNA | BC162345.1 | - | 1 - 2 | 754 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
