Assay Details
| Gene Symbol: tek | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131461.1 | NP_571536.1 | 4 - 5 | 679 | 78 | |
| XM_009301113.3 | XP_009299388.1 | 4 - 5 | 1228 | 78 | ||
| XM_021476003.1 | XP_021331678.1 | 4 - 5 | 1139 | 78 | ||
| XM_021476005.1 | XP_021331680.1 | 4 - 5 | 1136 | 78 | ||
| GenBank mRNA | AF053632.1 | - | 4 - 5 | 679 | 78 | |
| BC163579.1 | - | 4 - 5 | 679 | 78 | ||
More Information
Set Membership: |
|

tek
Genomic Map