Assay Details
| Gene Symbol: fbxw4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131521.2 | NP_571596.1 | 1 - 2 | 418 | 69 | |
| GenBank mRNA | AB022162.1 | - | 1 - 2 | 420 | 69 | |
| BC066481.1 | - | 1 - 2 | 322 | 69 | ||
More Information
Set Membership: |
|

fbxw4
Genomic Map