Assay Details
| Gene Symbol: hoxc12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131545.1 | NP_571620.1 | 1 - 2 | 615 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

hoxc12b| Gene Symbol: hoxc12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131545.1 | NP_571620.1 | 1 - 2 | 615 | 74 | |
Set Membership: |
|