Assay Details
Gene Symbol: nkx2.4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131589.1 | NP_571664.1 | 1 - 2 | 380 | 90 | |
GenBank mRNA | AF253054.1 | - | 1 - 2 | 380 | 90 | |
BC162296.1 | - | 1 - 2 | 377 | 90 | ||
BC162307.1 | - | 1 - 2 | 377 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
