Assay Details
| Gene Symbol: ybx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126457.1 | NP_001119929.1 | 7 - 8 | 1172 | 113 | |
| NM_131620.1 | NP_571695.1 | 7 - 8 | 1090 | 116 | ||
| GenBank mRNA | AF093129.1 | - | 7 - 8 | 1090 | 116 | |
| BC050156.1 | - | 7 - 8 | 1109 | 113 | ||
| BC070000.1 | - | 7 - 8 | 1090 | 113 | ||
| BC128885.1 | - | 7 - 8 | 1086 | 113 | ||
| BC154228.1 | - | 5 - 6 | 895 | 116 | ||
| BC160650.1 | - | 7 - 8 | 1096 | 116 | ||
More Information
Set Membership: |
|

ybx1
Genomic Map