Assay Details
Gene Symbol: p2rx3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131623.1 | NP_571698.1 | 4 - 5 | 511 | 106 | |
GenBank mRNA | AF237683.1 | - | 4 - 5 | 517 | 106 | |
AF238292.1 | - | 4 - 5 | 511 | 106 | ||
BC162279.1 | - | 4 - 5 | 497 | 106 | ||
BC162600.1 | - | 4 - 5 | 497 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
